Ergodyne™ Chill-Its™ 6665 Evaporative Cooling Vest
Lightweight, durable, and washable vest
Manufacturer: ERGODYNE 12535
 View more versions of this product
Catalog No. 19-154-642
 N/A
    N/A
Part Not Found 
    Specifications
    Length (English) 24 in.
    Length (Metric) 60.96cm
    Size XL
    Closure Type Zipper
    Color Hi-vis lime
    Product Type Chill-Its™ 6665 Evaporative Cooling Vest
    For Use With (Application) Construction, trades, maintenance, landscaping or grounds, assembly or fabrication, material handling, freight or baggage, warehousing or distribution, Iron/Steel fabrication
    Material Nylon
    Neck Style V-neck
    Quantity Each
    • Water-repellent liner and black cotton-poly elastic trim
    • Activated, polymer embedded fabric inside
    • Quilted nylon outer later
    • V-neck with zipper closure
    • Nylon
  • Description & Specifications
Related Products
Product Content Feedback