Learn More
Ergodyne™ Chill-Its™ 6700 Evaporative Cooling Tie Bandana
Cool comfort for 24 to 48 hours when activated in cool water
Manufacturer:  Ergodyne™ 12307
 View more versions of this product
Catalog No. 19-058-457
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Ideal for workers in construction, landscaping, foundries, mining, welding, factories, farming and food service.
  • Evaporative cooling works all day
  • Lightweight, comfrotable cotton construction provides all-day comfort
  • Super absorbent acrylic polymer activates easily when soaked in water for 15 mins.
  • Machine-washable; Reusable
Specifications
Specifications
Blue
Activated Polymers, tie closure
Chill-Its™ Evaporative Cooling Bandana
One size fits all
Ideal for workers in construction, landscaping, foundries, mining, welding, factories, farming and food service
Cotton
Safety