Ergodyne™ GloWear™ 8245PSV Public Safety Vests

Five point breakaway design

Manufacturer: ERGODYNE 23383

 View more versions of this product

Catalog No. 19-808-166

  Check Availability
Add to Cart

    Specifications

    No. of Pockets 1.0
    Size (Metric) Chest 91 to 112cm
    Tape Type 1 in. 3M™ Scotchlite™ reflective stripes
    Color Orange
    Size Small/Medium
    Size (English) Chest 36 to 44 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit