Ergodyne™ GloWear™ Hi-Vis Class II Vests
High visibility, glowing vests keep workers safe on the job
Manufacturer: ERGODYNE 21325
 View more versions of this product
Catalog No. 19-130-2305
  Check Availability
Add to Cart
    Specifications
    No. of Pockets 1.0
    Size (English) Chest 44 to 52 in.
    Size (Metric) Chest 112 to 132cm
    Tape Type 2 in. 3M™ Scotchlite™ reflective stripes
    Color Lime
    Material Polyester
    Size Large/X-Large
  • Description & Specifications
Related Products
Product Content Feedback