Ergodyne Knee Sleeves

Manufacturer: ERGODYNE 16505

 View more versions of this product

Catalog No. 19-037-835D

  Check Availability
Add to Cart

    Specifications

    Color Black
    Construction Single layer, Neoprene four-way stretch Fabric
    Design Contoured
    For Use With (Application) Provide glove-like fit and comfortable wear and provides uniform support and warmth to affected area
    Material Neoprene
    Model 600
    Size (English) 16 to 18 in.
    Size (Metric) 41 to 46cm
    Product Type Proflex™ Knee Sleeve
    Size XL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit