Learn More
Ergodyne™ ProFlex™ 1100SF Standard Spandex Back Support Brace
Manufacturer:  Ergodyne™ 11602
 View more versions of this product
Catalog No. 19-147-435
Add to cart
Description
Description
Specifications
Specifications
5.08 cm
Material handling, construction, warehousing/distribution, maintenance, freight/baggage, assembly/fabrication, landscaping/grounds, pall bearers
64 to 76 cm
Small
Black
Proflex™ Standard Back Support
Adjustable two stage
5 in.
280D Spandex
Patented non-conductive sticky fingers™ stays, 1.25 in. knit Elastic suspenders, power panel Positioners™, stretchable bias binding with zig-zag stitching
25 to 30 in.
1100SF