Ergodyne™ ProFlex™ 4000/4010 Wrist Supports

Flexible support that doesn't compromise productivity

Manufacturer: ERGODYNE 70016

 View more versions of this product

Catalog No. 19-035272E

  Check Availability
Add to Cart

    Specifications

    Closure Type Two stage hook and loop
    For Use With (Application) Food processing, picking, packing, assembly, fabrication and taffy pulling
    Product Type Proflex™ Single Strap Wrist Support
    Color Black
    Hand Left
    Includes Patented open-center Stay™, red line indicator, anti-bacterial lining
    Limitations Limits downward wrist movement
    Material Elastic
    Model 4000
    Size (English) 7 to 8 in.
    Size (Metric) 18 to 20cm
    Size Large

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit