Learn More
Ergodyne™ ProFlex™ 4020 Wrist Supports
Limit downward wrist movement during repetitive work
Manufacturer:  ERGODYNE 4020RTMD
 View more versions of this product
Catalog No. 19-808-100
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Two stage hook and loop
Contoured
Right
Limit downward wrist movement
4020
15.24 to 18cm
Medium
Gray
Designed to relieve stress associated with repetitive strain and overuse injuries and also for food processing, picking, packing, assembly, fabrication, office, retail and light industrial
Patented open-center Stay™, interior ventilation holes, anti-microbial lining
Neoprene
6 to 7 in.
Proflex™ Wrist Support