Ergodyne™ ProFlex Universal Wrist Wraps

Wrist wraps that provide support

Manufacturer: ERGODYNE 72234

 View more versions of this product

Catalog No. 19-037-832B

  Check Availability
Add to Cart

    Specifications

    Closure Type Hook and loop
    Color Tan
    For Use With (Application) Food processing, picking, packing, assembly, fabrication
    Hand Ambidextrous
    Includes Thumb loop
    Material Woven Elastic
    Model 420
    Size (English) 7 to 9 in.
    Size (Metric) 18 to 22.86cm
    Product Type Proflex™ Wrist Wrap
    Size Large/XL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit