Ergodyne Squids Glove Grabbers

Keeps gloves where you want them, when you want them

Manufacturer: ERGODYNE 19122

 View more versions of this product

Catalog No. 19-150-0004

  Check Availability
Add to Cart

    Specifications

    Includes Belt clip
    For Use With (Application) patented design opens smoother and wider and can hold gloves of every style and thickness
    Material Acetal Copolymer
    Size Universal
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit