Learn More
Sartorius Ergomate Pipet Accessories
Accessory for Ergomate Pipet
$88.60
Products 1
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
14-559-560
View Documents Promotion Details
Sartorius
783500
120mL reagent vessel 120mL
Case of 16 for $88.60
Add to cart
Description
Description
Ergomate removes the need to hold the plate at any angle and allows safe dispensing, resulting easier, safer and more precise pipetting.
According to ISO 8655 recommendations, dispensing especially small volumes should be done against the inside wall of receiving vessel. Ergomate is an ideal tool for finding proper positioning in pipetting.
Microplate grids for quick identification and sample management.
Grids are double sided. Reverse side featuring numbering from 1-12.
Plate Holder
When reversed plate holder creates an ideal aerosol barrier for use in hoods or on bench top surfaces.
Plate holder can be reversed and used for flat surface pipetting or as an incubation station with lid.
Ideal for safe transporting of plates.
Non autoclavable