Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERH Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318856PCP
Description
ERH Polyclonal antibody specifically detects ERH in Human samples. It is validated for ELISASpecifications
| ERH | |
| Polyclonal | |
| ELISA | |
| DROER, enhancer of rudimentary (Drosophila) homolog, enhancer of rudimentary homolog, enhancer of rudimentary homolog (Drosophila), FLJ27340 | |
| Produced in rabbits immunized with purified, recombinant Human ERH (Accession#: P84090; Met1-Lys104) | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 2079 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction