Alfa Aesar™ Erie Garnet, ca. 80% dye content

Manufacturer: Alfa Aesar™ A1421614

Quantity 25g

 View more versions of this product

Catalog No. AAA1421614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2429-70-1
    Molecular Formula C32H21N5O7S2Na2
    Formula Weight 697.65
    MDL Number MFCD00046442
    Synonym C.I. 22145; Congo Corinth
    Chemical Name or Material Erie Garnet, ≈80% dye content

    Specifications

    Formula Weight 697.65
    Molecular Formula C32H21N5O7S2Na2
    MDL Number MFCD00046442
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF eye irritation persists: Get medical advice/attention.
    Quantity 25g
    CAS 2429-70-1
    Synonym C.I. 22145; Congo Corinth
    Health Hazard 2 GHS H Statement
    H319
    Causes serious eye irritation.
    Chemical Name or Material Erie Garnet, ≈80% dye content

    GHS H Statement
    H319
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF eye irritation persists: Get medical advice/attention.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit