Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Erie Garnet, ca. 80% dye content

Manufacturer:  Alfa Aesar™ A1421622

Quantity 100g

 View more versions of this product

Catalog No. AAA1421622


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

2429-70-1
697.65
C.I. 22145; Congo Corinth
C32H21N5O7S2Na2
MFCD00046442
Erie Garnet, ≈80% dye content

Specifications

2429-70-1
697.65
C.I. 22145; Congo Corinth
Erie Garnet, ≈80% dye content
C32H21N5O7S2Na2
MFCD00046442
100g

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit