Learn More
Fisherbrand™ Erie Polyurethane Chair Chrome - High Bench Height with Chrome Foot Ring and Chrome Casters in Polyurethane Products for Science Education
Fisherbrand™ Erie Polyurethane Chairs in Chrome are height adjustable for use at 42 in. work surfaces and feature handle in the back rest, chrome adjustable foot ring, and self-braking casters. Self-skinning polyurethane is chemical, soil, and puncture resistant and easily cleaned.
$239.00 - $264.00
Specifications
Adjustable Yes
Base Style Polished Cast Aluminum
Material Polyurethane
Footrest Yes
With Armrest No
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S41165 Products for Science Education
View Documents
Blue
Each for $239.00
Add to cart
S36752 Products for Science Education
View Documents
Black
Each for $264.00
Add to cart
Description
Description
  • Adjustable from 23.5-33.5 inches from the top of the seat to the floor
  • Features manual back height and back depth adjustments
  • Built in handle for ease of movement
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
Polyurethane
No
Yes
High Bench Height
Polished Cast Aluminum
Yes
Yes
No