Learn More
Fisherbrand™ Erie Polyurethane Chair Chrome - Medium Bench Height with Adjustable Arms and Chrome Casters in Polyurethane
Fisherbrand™ Erie Polyurethane Chairs in Chrome are height adjustable for use at 36 in. work surfaces and feature handle in the back rest, adjustable arms, and self-braking casters. Self-skinning polyurethane is chemical, soil, and puncture resistant and easily cleaned.
$325.00 - $384.50
Specifications
Adjustable Yes
Material Polyurethane
Footrest No
With Armrest Yes
With Casters Yes
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-400-430
View Documents
Black
Each for $325.00
Add to cart
14-000-692
View Documents
Blue
Each for $384.50
Add to cart
Description
Description
  • Adjustable from 19.5-27 inches from the top of the seat to the floor
  • Features manual back height and back depth adjustments
  • Built in handle for ease of movement
  • 2-way adjustable arm rests adjust up and dow
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
No
Yes
Polyurethane
Yes
No