Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eriochrome Black T Indicator, 0.5% w/v in 2-Methoxyethanol, Reagents

Catalog No. 01335322
Click to view available options
Quantity:
100 mL
500 mL
1 L

Eriochrome Black T Indicator, 0.5% w/v in 2-Methoxyethanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Eriochrome Black T Indicator, 0.5% w/v in 2-Methoxyethanol
Boiling Point 123°C
CAS 109-86-4, 1787-61-7
Synonym Water hardness indicator
Percent Purity 0.5%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.97
Quantity 500 mL
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.