Eriochrome Black TS, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 29014

Packaging Glass amber
Quantity 120mL

 View more versions of this product

Catalog No. 2901-4

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 67-56-1,5470-11-1,1787-61-7
    CAS Min % 93.0,4.72,0.47
    CAS Max % 97.0,4.92,0.49
    Physical Form Liquid
    Chemical Name or Material Eriochrome Black, Test Solution

    Specifications

    Physical Form Liquid
    Color Clear dark red
    Quantity 120mL
    CAS Min % 93.0,4.72,0.47
    CAS 5470-11-1,1787-61-7,67-56-1
    CAS Max % 0.49,97.0,4.92
    Packaging Glass amber
    Chemical Name or Material Eriochrome Black, Test Solution

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit