Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eriochrome(R) Blue Black R, Spectrum™

Click to view available options
Quantity:
10 g
25 g
500 g
1 kg
Packaging:
Amber Glass Bottle
2538-85-4
2538-85-4
CAS Max % 1
Grade Reagent
Packaging Amber Glass Bottle
Quantity 500 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.