Learn More
Enzo Life SciencesSupplier Diversity Partner Erk1/2 ELISA kit (1x96wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI900152
The Erk1/2 EIA Kit is a complete assay for the quantitative determination of pan Erk1/2 concentration. . Alternative Name: MAPK2 3. MAP kinase 2 3. Sensitivity: 22.9 pg/ml (range 156 - 20,000 pg/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human, Mouse, Rat. Contents: Microtiter plate, Conjugate, Antibody, Assay buffer 21, Wash buffer concentrate, Standard, TMB Substrate, RIPA Cell lysis buffer 2, Stop solution 2
Catalog No. 50-658-179