Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK5/BMK1 Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI3H2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271243JF646

Catalog No. N271243J646


Add to cart

Description

Description

ERK5/BMK1 Monoclonal antibody specifically detects ERK5/BMK1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

ERK5/BMK1
OTI3H2
Flow Cytometry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
MAPK7
Full length recombinant protein of human ERK5/BMK1 (NP_002740) produced in HEK293T cells.
Immunogen affinity purified
Cell Cycle and Replication, Phospho Specific, Signal Transduction
Primary
5598.0
Flow Cytometry, Immunocytochemistry, Immunofluorescence
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Big MAP kinase 1, BMK-1, BMK1 kinase, BMK1EC 2.7.11.24, EC 2.7.11, ERK-5, ERK5MAPK 7, Extracellular signal-regulated kinase 5, extracellular-signal-regulated kinase 5, MAP kinase 7, mitogen-activated protein kinase 7, PRKM7ERK4
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit