Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  LIFE TECHNOLOGIES 720316

Encompass

Catalog No. 72-031-6


Add to cart

Description

Description

These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, Cat and Rat

ERK5 Polyclonal antibody specifically detects ERK5 in Human samples. It is validated for Immunocytochemistry, Immunofluorescence, Western Blot
Specifications

Specifications

ERK5
Immunocytochemistry, Immunofluorescence, Western Blot
0.5 mg/mL
PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4
Q13164
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5598
Liquid
Polyclonal
Unconjugated
MAPK7
big MAP kinase 1; Bmk1; BMK-1; BMK1 kinase; BNhp-1; BNhp1 kinase; ERK4; ERK5; ERK-5; Erk5-T; extracellular signal regulated kinase 5; extracellular signal-regulated kinase 5; extracellular-signal-regulated kinase 5; MAP kinase 7; MAPK 7; MAPK7; mitogen activated protein kinase 7; mitogen-activated protein kinase 7; PRKM7
Peptides corresponding to Human MAPK7 (aa 107-122, 500-519)
Affinity chromatography
RUO
Antibody
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit