Learn More
Eisco™ Erlenmeyer Flasks, Narrow Neck
Erlenmeyer flask made form high quality borosilicate glass. Flasks have a conical shape, narrow neck, and graduations. Conform to DIN ISO 1773 standard.
$2.00 - $69.00
Products 3
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
S27996 Products for Science Education
View Documents
Eisco™
CH0424C
50 mL
Each for $2.00
Add to cart
S15465
View Documents
Eisco™
CH0424D
100 mL
Each for $3.20
Add to cart
S40679 Products for Science Education
View Documents
Eisco™
CH0424K
500 mL
Each for $69.00
Add to cart
Description
Description
  • Erlenmeyer flask with graduations
  • Made from high quality borosilicate glass
  • Conical shape with narrow neck
  • As per DIN ISO 1773 standard