Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERO1L, Rabbit anti-Human,Mouse,Rat, Alexa Fluor 488, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB1002525AF488100UL

Catalog No. NB003450

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ERO1L Polyclonal antibody specifically detects ERO1L in Human, Mouse, Rat Samples. It is validated for Western Blot, Immunofluorescence.
Specifications

Specifications

ERO1L
Alexa Fluor 488
50mM Sodium Borate
EC 1.8.4, EC 1.8.4.-, Endoplasmic oxidoreductin-1-like protein, ERO1 (S. cerevisiae)-like, ERO1A, ERO1-alpha, ERO1-L, ERO1-L-alpha, ERO1-like (S. cerevisiae), ERO1-like protein alpha, oxidoreductin-1-L-alpha
Rabbit
IgG
100 uL
Cancer, ER Markers, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction
Primary
30001
Western Blot, Immunofluorescence
Western Blot, Immunocytochemistry/Immunofluorescence
Affinity Purified
ERO1A
A synthetic peptide made to the N-terminal region of human ERO1L [UniProt Q96HE7]
Immunogen affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human, Mouse, Rat
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit