Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERO1LB Mouse anti-Human, Janelia Fluor 646, Clone: M37-P5D11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP250389JF646

Catalog No. NBP50389646


Add to cart

Description

Description

ERO1LB Monoclonal antibody specifically detects ERO1LB in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

ERO1LB
M37-P5D11
Immunohistochemistry
Purified
ERO1B
Peptide - LAPSRGEDDG
Protein G purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
DKFZp779C1042, DKFZp779I0141, EC 1.8.4, EC 1.8.4.-, Endoplasmic oxidoreductin-1-like protein B, ERO1-L(beta), ERO1-L-beta, ERO1-like beta (S. cerevisiae), ERO1-like protein beta, FLJ11003, oxidoreductin-1-L-beta
Mouse
IgG1 Kappa
0.1mL
Primary
56605
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit