Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERp57/PDIA3 Mouse anti-Bovine, Canine, Guinea Pig, Hamster, Human, Monkey, Mouse, Porcine, Rabbit, Rat, Clone: Map.ERp57 (GRP58), Novus Biologicals™

Mouse MonoclonalAntibody

$229.00 - $351.00

Specifications

Antigen ERp57/PDIA3
Clone Map.ERp57 (GRP58)
Dilution Western Blot 1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
NB59689200U View Documents Novus BiologicalsSupplier Diversity Partner
NBP259689200UG
200 ug Unconjugated Each for $351.00
Add to cart
 
NB5968950UG View Documents Novus BiologicalsSupplier Diversity Partner
NBP25968950UG
50 ug Unconjugated Each for $229.00
Add to cart
 
Description

Description

ERp57/PDIA3 Monoclonal antibody specifically detects ERp57/PDIA3 in Human, Mouse, Rat, Porcine, Bovine, Canine, Guinea Pig, Hamster, Monkey, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

ERp57/PDIA3
Western Blot 1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100
Purified
Human, Mouse, Rat, Porcine, Bovine, Canine, Guinea Pig, Hamster, Monkey, Rabbit
PBS pH7.4, 50% glycerol with 0.09% Sodium Azide
Human recombinant ERp57 (Grp58)
Monoclonal
Protein G purified
Map.ERp57 (GRP58)
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Mouse
RUO
2923
IgG1
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit