Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERP72 Mouse anti-Human, Mouse, Clone: OTI2A11, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525769

Catalog No. PIMA525769


Add to cart

Description

Description

ERp72 is an endoplasmic reticulum luminal protein that is both a stress protein and a member of the protein disulfide isomerase family of proteins. Sequence analysis of ERp72 shows that it shares sequence identity with PDI (protein disulphide isomerase) at three discrete regions, as well as containing the ER retention signal KEEL at its C-terminus. ERp72 is shown to have three copies of sequence believed to be CGHC-containing active sites, which are also found in PDI. These CGHC sites are critical sites that are involved in the protein disulphide isomerization process. These results suggest that ERp72 is a newly described member of the family of CGHC-containing proteins that functions as a molecular chaperone.
Specifications

Specifications

ERP72
Monoclonal
0.99 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P13667, P08003
ERP70, ERP72, ERp-72, ER protein 70, ER protein 72, endoplasmic reticulum resident protein 70, endoplasmic reticulum resident protein 72, protein disulfide isomerase related protein (calcium-binding protein, intestinal-related), protein disulfide isomerase-associated 4, protein disu
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
12304, 9601
Immunohistochemistry (Paraffin), Western Blot
OTI2A11
Unconjugated
PDIA4
Liquid
PDIA4
Full length human recombit protein of PDIA4 produce in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit