Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse/Rat ESRRB, Synthetic Peptide, Invitrogen™
Synthetic Peptide
Supplier: Thermo Scientific PEP251
Description
ERR beta/ESRRB Synthetic Peptide, PEP-251, from Invitrogen™.
Estrogen-related receptor 2 (ERR beta, ERR2) is an orphan member of the nuclear hormone receptor superfamily. All the ERR family members share an almost identical DNA-binding domain, which has 68% amino acid identity with that of estrogen receptor. It has been shown the ERR2 plays a critical functional role in placental development. This peptide corresponds to amino acids D(376) YELSQRHEEPRR (388) of mouse and rat ERR2 and can be used as a blocking peptide with polyclonal antibody PA1-315.Specifications
26380, 299210 | |
purified | |
Mouse/Rat ESRRB | |
-20°C | |
Lyophilized | |
Blocking Assay, Control | |
Esrrb | |
No preservative | |
50 μg |
Mouse, Rat | |
P11475, Q61539 | |
1 mg/mL | |
Synthetic Peptide | |
water with no preservative | |
Steroid hormone receptor ERR2, Estrogen-related receptor beta, ERR-beta, Estrogen receptor-like 2, ERR beta-2 | |
Yes | |
Peptide | |
RUO - research use only |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction