Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERR gamma/NR3B3 Mouse anti-Human, DyLight™ 650, Clone: OTI1E5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270691C

Catalog No. NBP270691C


Add to cart

Description

Description

ERR gamma/NR3B3 Monoclonal antibody specifically detects ERR gamma/NR3B3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

ERR gamma/NR3B3
OTI1E5
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
ESRRG
Recombinant protein fragment corresponding to amino acids 24-263 of human ERR gamma/NR3B3 (NP_001429) produced in E.coli.
Immunogen affinity purified
GPCR
Primary
2104.0
Immunohistochemistry, Western Blot
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
DKFZp781L1617, ERR gamma-2, ERR3, ERRG2, ERRgamma, Estrogen receptor-related protein 3, estrogen-related receptor gamma, FLJ16023, KIAA0832, NR3B3, Nuclear receptor subfamily 3 group B member 3
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit