Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERR gamma/NR3B3 Antibody (OTI1E5), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270691JF646
Description
ERR gamma/NR3B3 Monoclonal specifically detects ERR gamma/NR3B3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ERR gamma/NR3B3 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ESRRG | |
Recombinant protein fragment corresponding to amino acids 24-263 of human ERR gamma/NR3B3 (NP_001429) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1E5 | |
Western Blot, Immunohistochemistry | |
DKFZp781L1617, ERR gamma-2, ERR3, ERRG2, ERRgamma, Estrogen receptor-related protein 3, estrogen-related receptor gamma, FLJ16023, KIAA0832, NR3B3, Nuclear receptor subfamily 3 group B member 3 | |
Mouse | |
Affinity Purified | |
GPCR | |
2104.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction