Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ertapenem sodium, 85%, Acros Organics™

Click to view available options
Quantity:
250mg
1g
153773-82-1
C22H24N3NaO7S
497.5
ZXNAQFZBWUNWJM-HRXMHBOMSA-M
1-azabicyclo 3.2.0 hept-2-ene-2-carboxylic acid, 3-3s,5s-5-3-ca rboxyphenyl amino carbonyl-3-pyrrolidinyl thio-6-1r-1-hydroxyethy l-4-methyl-7-oxo-, monosodium salt, 4r,5s,6s
131675376
CHEBI:60070
sodium;(2S,4S)-4-[[(4R,5S,6S)-2-carboxy-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-en-3-yl]sulfanyl]-N-(3-carboxyphenyl)pyrrolidine-2-carboximidate
CC1C2C(C(=O)N2C(=C1SC3CC(NC3)C(=NC4=CC=CC(=C4)C(=O)O)[O-])C(=O)O)C(C)O.[Na+]
153773-82-1
C22H24N3NaO7S
497.5
ZXNAQFZBWUNWJM-HRXMHBOMSA-M
1-azabicyclo 3.2.0 hept-2-ene-2-carboxylic acid, 3-3s,5s-5-3-ca rboxyphenyl amino carbonyl-3-pyrrolidinyl thio-6-1r-1-hydroxyethy l-4-methyl-7-oxo-, monosodium salt, 4r,5s,6s
131675376
CHEBI:60070
sodium;(2S,4S)-4-[[(4R,5S,6S)-2-carboxy-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-en-3-yl]sulfanyl]-N-(3-carboxyphenyl)pyrrolidine-2-carboximidate
CC1C2C(C(=O)N2C(=C1SC3CC(NC3)C(=NC4=CC=CC(=C4)C(=O)O)[O-])C(=O)O)C(C)O.[Na+]
Packaging Glass Bottle
Quantity 1g
Melting Point 230.0°C to 234.0°C
Loss on Drying 15% to 19%
Infrared Spectrum Authentic
Assay Percent Range 80% min. (HPLC)
Beilstein 15,3731
Formula Weight 497.5
Percent Purity 85%
Chemical Name or Material Ertapenem sodium

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air

GHS Signal Word: Warning