Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ertapenem sodium salt, 100mg/mL in distilled water, sterile-filtered, Alfa Aesar™

Click to view available options
Quantity:
1mL
5 x 1mL
153773-82-1
C22H24N3NaO7S
497.498
MFCD09475566
ZXNAQFZBWUNWJM-HRXMHBOMSA-M
131675376
CHEBI:60070
sodium;(2S,4S)-4-[[(4R,5S,6S)-2-carboxy-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-en-3-yl]sulfanyl]-N-(3-carboxyphenyl)pyrrolidine-2-carboximidate
CC1C2C(C(=O)N2C(=C1SC3CC(NC3)C(=NC4=CC=CC(=C4)C(=O)O)[O-])C(=O)O)C(C)O.[Na+]
153773-82-1
C22H24N3NaO7S
497.498
MFCD09475566
ZXNAQFZBWUNWJM-HRXMHBOMSA-M
131675376
CHEBI:60070
sodium;(2S,4S)-4-[[(4R,5S,6S)-2-carboxy-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-en-3-yl]sulfanyl]-N-(3-carboxyphenyl)pyrrolidine-2-carboximidate
CC1C2C(C(=O)N2C(=C1SC3CC(NC3)C(=NC4=CC=CC(=C4)C(=O)O)[O-])C(=O)O)C(C)O.[Na+]
Applications
Ertapenem has in vitro activity against gram-positive and gram-negative aerobic and anaerobic bacteria. The bactericidal activity of ertapenem results from the inhibition of cell wall synthesis and is mediated through ertapenem binding to penicillin binding proteins.

Solubility
Fully miscible in water. Soluble in DMSO.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Physical Form Liquid
Concentration 100mg/mL in distilled water
Quantity 5 x 1mL
Chemical Name or Material Ertapenem sodium salt, sterile-filtered

storageNote1 : −30° to −10°C