Ertco™ Exact-Temp™ Mercury Thermometers
For total immersion
Mounted in shatterproof plastic bottle filled with liquid to ensure accurate readings despite opening and closing of instrument door.
Specifications
Certifications NIST
View More Specs
 Alert
The products indicated contain mercury. Do not put products containing mercury in the trash. They should be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
  • Products 4
  • Description & Specifications
Products
Catalog Number Mfr. No. Metric Temperature Range Metric Length Subdivisions For Use With Application Price Quantity & Availability    
15059143
LUDWIG SCHNEIDER
I-030-1
+18° to +50°C 135mm 0.5°C Incubator N/A N/A N/A  
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
15059144
LUDWIG SCHNEIDER
OV-070
20° to 130°C 135mm 1°C Oven N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
15059141
LUDWIG SCHNEIDER
F-010-1
-30° to 0°C 115mm 0.5°C Freezer N/A N/A N/A
 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
15059142
LUDWIG SCHNEIDER
R-020-1
-5° to +15°C 125mm 0.5°C Refrigerator N/A N/A N/A
 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
Descriptions & Specifications
Related Products