Learn More
Ertco™ Precision-Grade Armored Thermometers
Provide accurate temperature readings while avoiding the environmental hazards associated with the disposal of mercury
Manufacturer:  *LUDWIG SCHNEIDER 11354/ARM12
 View more versions of this product
Catalog No. 15-060-283
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
NIST
12 in.
1°C
Organic
Precision
305mm
-10° to +200°C