Learn More
Ertco Precision standard taper Joint Thermometers
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
15-059-317
|
Ertco
ST03150SC |
N/A | 10deg. to 150deg. C; Contains Mercury!Contains Mercury!; Immersion: 75mm |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
With 10/30 standard taper ground joints fused to the stem at the immersion point. Accurate to within NIST tolerances. Yellow back. 1°C divisions. Coated with PTFE resin except for the ground joint. Protects against shock as well as contamination in case of breakage.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.