Ertco™ PTFE Coated Glass Special-Application Red-Spirit™-Filled Thermometers
Individually serialized
Specifications
Material PTFE Coated Glass
View More Specs
  • Products 2
  • Description & Specifications
Products
Catalog Number Mfr. No. Metric Temperature Range Subdivisions For Use With Application Immersion Length Price Quantity & Availability    
15-060-253
Thermo Scientific
10970-05
+35° to +200°C 1°C Vacuum Ovens Total N/A N/A N/A
 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
15-060-237
Thermo Scientific
10919-05
25°/37°/56°C 0.5°C Incubator, Water Bath 76mm N/A N/A N/A
 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View Substitute Products
Descriptions & Specifications
Related Products