Ertco™ standard taper Ground Joint Thermometers
10/30 Standard Taper
Manufacturer: *LUDWIG SCHNEIDER 10948/BF
 View all options for this product
Catalog No. 15-002E
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    • Suitable for high-vacuum work
    • Precisely ground to a standard taper
    • Taper ensures true interchangeability with female taper of similar quality and suitable size
    • Engraved stem; yellow back; 1°C divisions
    • Graduations and numbers are etched on glass
  • Description & Specifications
Related Products
Product Content Feedback