Learn More
Ertco™ Universal Angle Thermometers
Dial angle can be adjusted for easy reading
Manufacturer: Ertco™ MA3004006
Description
- Dual scale
- Have all the features of the Ertco Back-Connected Thermometers, plus the dial angle can be adjusted for easy reading
- Simple adjustment of the stainless-steel harness allows case rotation of 360° and stem variation of more than 180°
- With 1/2 NPT connections