Learn More
Ertco™ Universal Angle Thermometers
Dial angle can be adjusted for easy reading
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
15-060-128
View Documents
Ertco™
MA3004006
N/A Thrm Bi 3 in. /4 in. 0/250f -10/120c This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Dual scale
  • Have all the features of the Ertco Back-Connected Thermometers, plus the dial angle can be adjusted for easy reading
  • Simple adjustment of the stainless-steel harness allows case rotation of 360° and stem variation of more than 180°
  • With 1/2 NPT connections