Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERVK-9 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549533

Catalog No. PA549533

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Retroviral envelope proteins mediate receptor recognition and membrane fusion during early infection. Endogenous envelope proteins may have kept, lost or modified their original function during evolution. This endogenous envelope protein has lost its original fusogenic properties.
Specifications

Specifications

ERVK-9
Polyclonal
PBS with 0.09% sodium azide
P63128
Endogenous retrovirus group K member 9 Env polyprotein, EnvK4 protein, Envelope polyprotein, HERV-K(C6) envelope protein, HERV-K109 envelope protein, HERV-K_6q14.1 provirus ancestral Env polyprotein, Surface protein, SU, Transmembrane protein, TM, ERVK-9
Rabbit
IgG
50 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100616102
Western Blot
Unconjugated
ERVK-9
Liquid
ERVK-9
KLH conjugated synthetic peptide between 636-669 amino acids from human ERVK-9.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit