Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythorbic Acid, FCC, 99-100.5%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
89-65-6
C6H7NaO6
198.11
IFVCRSPJFHGFCG-HXPAKLQESA-N
sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
[Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O
89-65-6
C6H7NaO6
198.11
IFVCRSPJFHGFCG-HXPAKLQESA-N
sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
[Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

E1004, 89-65-6, C6H8O6

Spectrum™ Erythorbic Acid, FCC is used as a food preservative additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.003
Loss on Drying 0.004
Quantity 100 g
Optical Rotation -16.5° to -18.0°
CAS Max % 1
Percent Purity 99 to 100.5%