Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythritol, FCC, 99.5-100.5%, Spectrum™ Chemical
SDP

Catalog Number 18602445
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
149-32-6
C4H10O4
122.12
MFCD00004710
UNXHWFMMPAWVPI-UHFFFAOYNA-N
butane-1,2,3,4-tetrol
OCC(O)C(O)CO
149-32-6
C4H10O4
122.12
MFCD00004710
UNXHWFMMPAWVPI-UHFFFAOYNA-N
butane-1,2,3,4-tetrol
OCC(O)C(O)CO

E1494, 149-32-6, C4H10O4

Spectrum™ Chemical Erythritol, FCC is a sugar alcohol alternative sweetener. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Loss on Drying 0.2%
Packaging Poly Pail
Quantity 5 kg
Percent Purity 99.5 to 100.5%
Grade FCC
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit