Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-erythro-Aleuritic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
533-87-9
C16H32O5
304.41
MFCD00135596
MEHUJCGAYMDLEL-LSDHHAIUSA-N
aleuretic acid,8,9,15-trihydroxypentadecane-1-carboxylic acid,9,10,16-trihydroxypalmitic acid,10r,9s-9,10,16-trihydroxyhexadecanoic acid,9s,10r-9,10,16-trihydroxyhexadecanoic acid,unii-442lq2k03a component
7269316
(9S,10R)-9,10,16-trihydroxyhexadecanoic acid
C(CCCC(C(CCCCCCO)O)O)CCCC(=O)O
533-87-9
C16H32O5
304.41
MFCD00135596
MEHUJCGAYMDLEL-LSDHHAIUSA-N
aleuretic acid,8,9,15-trihydroxypentadecane-1-carboxylic acid,9,10,16-trihydroxypalmitic acid,10r,9s-9,10,16-trihydroxyhexadecanoic acid,9s,10r-9,10,16-trihydroxyhexadecanoic acid,unii-442lq2k03a component
7269316
(9S,10R)-9,10,16-trihydroxyhexadecanoic acid
C(CCCC(C(CCCCCCO)O)O)CCCC(=O)O
Melting Point 100°C to 105°C
Quantity 25g
Color White to Beige
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula HO(CH2)5CH(OH)CH(OH)(CH2)7COOH
Beilstein 03, III, 901
Merck Index 15, 225
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,ethyl acetate,,soluble in aq. alkali solution
Formula Weight 304.41
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material (±)-erythro-Aleuritic acid, 95%
Show More Show Less

EINECSNumber : 208-578-8

RTECSNumber : ML9850000

TSCA : TSCA