Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythroid Cells Mouse anti-Rat, FITC, Clone: HIS49, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 11975582

Catalog No. 50-112-2185


Add to cart

Description

Description

Description: The monoclonal antibody HIS49 reacts with an unknown antigen expressed by rat erythroid cells and their progenitors. HIS49 does not react with lymphocytes, monocytes or granulocytes. Applications Reported: This HIS49 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HIS49 antibody has been tested by flow cytometric analysis of rat bone marrow cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 µm post-manufacturing filtered.

Specifications

Specifications

Erythroid Cells
Monoclonal
0.5 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Erythrocytes
IgM, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
Rat
Flow Cytometry
HIS49
FITC
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.