Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythromycin Estolate, USP, 600 μg/g, Spectrum™

Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum

E1304, 3521-62-8, C40H71NO14•C12H26O4S

Spectrum™ Erythromycin Estolate, USP , a common antibiotic, can be used to treat several types of bacterial infections and as an alternative treatment to treat sexually transmitted diseases. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS 3521-62-8
CAS Max % 1
Molecular Formula C52H97NO18S
Packaging Fiber Drum
InChI Key AWMFUEJKWXESNL-JZBHMOKNSA-N
SMILES CCCCCCCCCCCCOS(O)(=O)=O.CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2OC(=O)CC)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O
IUPAC Name (2S,3R,4S,6R)-4-(dimethylamino)-2-{[(3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-14-ethyl-7,12,13-trihydroxy-4-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,7,9,11,13-hexamethyl-2,10-dioxo-1-oxacyclotetradecan-6-yl]oxy}-6-methyloxan-3-yl propanoate; (dodecyloxy)sulfonic acid
Molecular Weight (g/mol) 1056.40
Water 0.04
Concentration 600 μg/g
Quantity 25 kg
Show More Show Less