Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythropoietin alpha, human, Alfa Aesar™

Click to view available options
Quantity:
2kU
500 units
11096-26-7
C22H22ClKN6O
461.007
OXCMYAYHXIHQOA-UHFFFAOYSA-N
Recombinant Human Erythropoietin-^a,rHuEPO-^a
11751549
potassium;[2-butyl-5-chloro-3-[[4-[2-(1,2,3-triaza-4-azanidacyclopenta-2,5-dien-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol
CCCCC1=NC(=C(N1CC2=CC=C(C=C2)C3=CC=CC=C3C4=NN=N[N-]4)CO)Cl.[K+]
11096-26-7
C22H22ClKN6O
461.007
OXCMYAYHXIHQOA-UHFFFAOYSA-N
Recombinant Human Erythropoietin-^a,rHuEPO-^a
11751549
potassium;[2-butyl-5-chloro-3-[[4-[2-(1,2,3-triaza-4-azanidacyclopenta-2,5-dien-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol
CCCCC1=NC(=C(N1CC2=CC=C(C=C2)C3=CC=CC=C3C4=NN=N[N-]4)CO)Cl.[K+]
Packaging Plastic bottle
Quantity 500 units
Formula Weight 21 kd
Physical Form Lyophilized Powder
Chemical Name or Material Erythropoietin alpha