Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythropoietin alpha, human, Thermo Scientific Chemicals

Catalog Number p-7076739
Click to view available options
Quantity:
2 kU
500 U
11096-26-7
C22H22ClKN6O
461.007
OXCMYAYHXIHQOA-UHFFFAOYSA-N
Recombinant Human Erythropoietin-^a,rHuEPO-^a
11751549
potassium;[2-butyl-5-chloro-3-[[4-[2-(1,2,3-triaza-4-azanidacyclopenta-2,5-dien-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol
CCCCC1=NC(=C(N1CC2=CC=C(C=C2)C3=CC=CC=C3C4=NN=N[N-]4)CO)Cl.[K+]
11096-26-7
C22H22ClKN6O
461.007
OXCMYAYHXIHQOA-UHFFFAOYSA-N
Recombinant Human Erythropoietin-^a,rHuEPO-^a
11751549
potassium;[2-butyl-5-chloro-3-[[4-[2-(1,2,3-triaza-4-azanidacyclopenta-2,5-dien-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol
CCCCC1=NC(=C(N1CC2=CC=C(C=C2)C3=CC=CC=C3C4=NN=N[N-]4)CO)Cl.[K+]

Used in the study of red blood cell growth and differentiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Packaging Plastic bottle
Quantity 2 kU
Formula Weight 21 kDa
Physical Form Lyophilized Powder
Chemical Name or Material Erythropoietin alpha
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit