Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythropoietin Mouse anti-Human, Clone: EPO/1367- Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

$435.85

Specifications

Antigen Erythropoietin
Clone EPO/1367
Host Species Mouse
Isotype IgG
Gene Alias EP, epoetin, erythropoietin, MGC138142, MVCD2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB254447100 View Documents Novus Biologicals
NBP254447100UG
100 ug Each for $435.85
Add to cart
 
NB254447200 View Documents Novus Biologicals
NBP254447200UG
200 ug This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Erythropoietin Monoclonal antibody specifically detects Erythropoietin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications

Specifications

Erythropoietin
Mouse
EP, epoetin, erythropoietin, MGC138142, MVCD2
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Store at -20 to -80C. Avoid freeze-thaw cycles.
Monoclonal
1 mg/mL
EPO/1367
IgG
Human
RUO
Purified
Primary
PBS with No Preservative

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit