Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erythropoietin/EPO Mouse anti-Human, DyLight 405, Clone: Epo1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB11016997V

Catalog No. NB11016997V

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Erythropoietin/EPO Monoclonal antibody specifically detects Erythropoietin/EPO in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Erythropoietin/EPO
Epo1
Western Blot, ELISA
Purified
EPO
Recombinant full length protein (Human).
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
EPO (Epo1)
Western Blot, ELISA
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Mouse
IgG1
0.2 ml
Angiogenesis, Cancer, Cytokine Research, HIF Target Genes, Hypoxia
Primary
2056
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit