Learn More
FUJIFILM Wako PYROSTAR™ ES-F 2 mL Bulk Kit
Endotoxin-specific and specially formulated to be unreactive to (1,3)-ß-D-glucan, Dual-purpose formulated to be used as gel clot or kinetic turbidimetric assay, Bulk configuration for large volume labs
Supplier: FUJIFILM Wako WPEM20003
Description
- Endotoxin-specific reagent avoids false positive results from glucans
- Dual-purpose reagent can perform either gel clot or kinetic turbidimetric assay (KTA)
- Kinetic turbidimetric sensitivities range from 0.001 to 10 EU/mL
- Due to high sensitivity, chances of interference are reduced
- Facilitates easier transition from gel clot to KTA
Specifications
Lyophilized | |
100 x 2 mL vials of lysate | |
0.03 EU/mL |
2 mL | |
100 Vials |